Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

557595

Sigma-Aldrich

10-(2-Benzothiazolyl)-2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-1H,5H,11H-(1)benzopyropyrano(6,7-8-I,j)quinolizin-11-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N2O2S
CAS Number:
Molecular Weight:
430.56
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

mp

230-232 °C (lit.)

λmax

478 nm in chloroform

SMILES string

CC1(C)CCN2CCC(C)(C)c3c4OC(=O)C(=Cc4cc1c23)c5nc6ccccc6s5

Application

Green dopant for OLED applications

Features and Benefits

Fluorescent dye
This sterically hindered coumarine dopant is an excellent choice for a green dopant for use in passive-matrix OLEDs.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen, C.H.; Tang, C.W.
Applied Physics Letters, 79, 3712-3712 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service