Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55752

Supelco

2-Ethoxynaphthalene

analytical standard

Synonym(s):

2-naphthol ethyl ether, Ethyl 2-naphthyl ether, Neroline, beta-naphthol ethyl ether, beta-naphthyl ethyl ether, bromelia, ethyl beta-naphtholate, ethyl beta-naphthyl ether, nerolin bromelia

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O
CAS Number:
Molecular Weight:
172.22
Beilstein:
2042397
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

35-37 °C (lit.)

solubility

alcohol: soluble
dipropylene glycol: soluble
isopropyl myristate: soluble

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOc1ccc2ccccc2c1

InChI

1S/C12H12O/c1-2-13-12-8-7-10-5-3-4-6-11(10)9-12/h3-9H,2H2,1H3

InChI key

GUMOJENFFHZAFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
The fluorescence of 2-Ethoxynaphthalene has been shown to be weakly quenched by m-dimethlphthalate and methyl benzoate. Importantly, at increased concentration methyl benzoate enhances the fluorescence by increasing the polarity of the solvent .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service