Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55510

Sigma-Aldrich

N-Hydroxymaleimide

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3NO3
CAS Number:
Molecular Weight:
113.07
Beilstein:
1524594
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥97.0% (HPLC)

mp

130-133 °C (dec.) (lit.)
130-133 °C

SMILES string

ON1C(=O)C=CC1=O

InChI

1S/C4H3NO3/c6-3-1-2-4(7)5(3)8/h1-2,8H

InChI key

BUXKULRFRATXSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the preparation of N-hydroxysuccinimid ester polymers via polymerizable active esters

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y.A. Davidovitch et al.
Acta Chemica Scandinavica, B 33, 311-311 (1979)
M. Akiyama et al.
Journal of the Chemical Society. Perkin Transactions 1, 2122-2122 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service