Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54402

Sigma-Aldrich

2-(Methylsulfonyl)ethanol

purum, ≥98.0% (GC)

Synonym(s):

2-Hydroxyethyl methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2CH2CH2OH
CAS Number:
Molecular Weight:
124.16
Beilstein:
1746915
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% (GC)

bp

148-149 °C (lit.)

mp

31-33 °C (lit.)

storage temp.

2-8°C

SMILES string

CS(=O)(=O)CCO

InChI

1S/C3H8O3S/c1-7(5,6)3-2-4/h4H,2-3H2,1H3

InChI key

KFTYFTKODBWKOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Methylsulfonyl)ethanol is an asymmetric sulfone.

Application

2-(Methylsulfonyl)ethanol may be used as a reagent for the synthesis of electron deficient phenols from the corresponding fluorides.[1] It may also be used in the preparation of 2-cyanophenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild conversion of electron deficient aryl fluorides to phenols using 2-(methylsulfonyl) ethanol.
Rogers JF and Green DF.
Tetrahedron Letters, 43(19), 3585-3587 (2002)
S G Cohen et al.
Journal of medicinal chemistry, 28(9), 1309-1313 (1985-09-01)
Reversible inhibitors for acetylcholinesterase, AcChE, have been studied. Sterically similar alcohols with tetra-substituted uncharged beta groups, (CH3)3SiCH2CH2OH (I), (CH3)3CCH2CH2OH (IA), and CH3S(O2)CH2CH2OH (VII), bind similarly, KI = 3-9 mM, and each binds similarly to its acetate substrate; cationic analogues, (CH3)3N+CH2CH2OH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service