531340
8-Amino-1-naphthalenesulfonic acid
Synonym(s):
1-Amino-8-naphthalenesulfonic acid, Peri acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
>350 °C (lit.)
SMILES string
Nc1cccc2cccc(c12)S(O)(=O)=O
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biochemistry and molecular biology international, 32(6), 1121-1127 (1994-04-01)
We have studied the importance of hydrophobic interactions in the fusion activity of Sendai virus. Viral fusion activity towards model lipid membranes was monitored using a fluorescence dequenching assay. Fusion was dependent on the composition of the target membrane and
Biochimica et biophysica acta, 1022(2), 171-180 (1990-02-28)
The molecular mechanism of the solubilisation of phospholipid bilayers by nonionic detergents was studied by turbidity changes, carboxyfluorescein fluorescence dequenching, steady-state and time-resolved fluorescence anisotropy of DPH, lifetime measurements, ANS binding and 31P-NMR. Particular attention has been paid to the
Analytical biochemistry, 185(2), 220-229 (1990-03-01)
Reaction of FMNH2 and O2 with bacterial luciferase followed by blue light irradiation results in a product previously claimed to have the same fluorescence spectral distribution as the bioluminescence. Preparations of this "high fluorescence" intermediate, however, contain two fluorescent components
Archives of biochemistry and biophysics, 302(1), 128-133 (1993-04-01)
Recombinant glutathione S-transferase P (GST-P) was purified in a homogeneous state. Fatty acid analysis of the enzyme revealed that the final enzyme preparation endogenously bound fatty acids, mostly palmitic acid or stearic acid, which were difficult to dissociate from the
Organic & biomolecular chemistry, 5(22), 3637-3650 (2007-11-01)
The design, synthesis, and characterization of novel cavitand-based hetero-TASPs, TASPs having different peptide sequences within one bundle, are described. Three families of hetero-TASPs were designed: the LG3/LG2 family (different linker lengths), LG3/AG3 family (altering helix hydrophobicity), and the LG3/LG2C family
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service