Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52288

Supelco

6-Methoxy-2-naphthol

analytical standard

Synonym(s):

2-Hydroxy-6-methoxynaphthalene, 6-Methoxynaphthalen-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
Beilstein:
2043873
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

form

solid

shelf life

limited shelf life, expiry date on the label

SMILES string

COc1ccc2cc(O)ccc2c1

InChI

1S/C11H10O2/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7,12H,1H3

InChI key

WWPKRXOOVICNJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service