Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51299

Supelco

Propionaldehyde, DAIH derivative

analytical standard

Synonym(s):

2-Diphenylacetyl-3-(propylidene-hydrazono)indan-1-one, 2-Diphenylacetyl-indan-1,3-dione-1-propylidenehydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22N2O2
CAS Number:
Molecular Weight:
394.47
Beilstein:
2019900
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

CC\C=N\N=C1/C(C(=O)C(c2ccccc2)c3ccccc3)C(=O)c4ccccc14

InChI

1S/C26H22N2O2/c1-2-17-27-28-24-20-15-9-10-16-21(20)25(29)23(24)26(30)22(18-11-5-3-6-12-18)19-13-7-4-8-14-19/h3-17,22-23H,2H2,1H3/b27-17+,28-24-

InChI key

WYNULXLCEYYXEZ-YACXLVKKSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service