Skip to Content
MilliporeSigma
All Photos(1)

Documents

49868

Supelco

2′-Aminoacetophenone

analytical standard

Synonym(s):

2-Acetylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4COCH3
CAS Number:
Molecular Weight:
135.16
Beilstein:
386122
EC Number:
MDL number:
UNSPSC Code:
12164502

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.613-1.619
n20/D 1.614 (lit.)

bp

85-90 °C/0.5 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

storage temp.

2-8°C

SMILES string

CC(=O)c1ccccc1N

InChI

1S/C8H9NO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,9H2,1H3

InChI key

GTDQGKWDWVUKTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′-Aminoacetophenone is a volatile aromatic compound, which is typically used as a key flavor ingredient in food and beverages.

Application

2′-Aminoacetophenone may be used as an analytical standard for the determination of the analyte in livestock particulate matter, and grape-derived beverages by gas chromatography-mass spectrometry-olfactometry (GC-MS-O) and GC-MS, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of volatile organic compounds and odorants associated with swine barn particulate matter using solid-phase microextraction and gas chromatography-mass spectrometry-olfactometry
Cai L, et al.
Journal of Chromatography A, 1102(1-2), 60-72 (2006)
2-Aminoacetophenone as a potential breath biomarker for Pseudomonas aeruginosa in the cystic fibrosis lung
Scott-Thomas AJ, et al.
BMC Pulmonary Medicine, 10(1), 56-56 (2010)
Rapid determination of the aromatic compounds methyl-anthranilate, 2?-aminoacetophenone and furaneol by GC-MS: Method validation and characterization of grape derivatives
Dutra M, et al.
Food Research International, 107(1-2), 613-618 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service