Skip to Content
MilliporeSigma
All Photos(1)

Documents

49500

Supelco

Sieve Standard, traceable to NIST SRM

63 μm (230 mesh)

Synonym(s):

Sieve calibration standard, 63 μm (230 mesh)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

form

particles

manufacturer/tradename

Whitehouse Scientific Ltd SS397

particle size

63 μm (230 mesh)

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Sieve Standard, traceable to NIST SRM is ideal for profiling particle size distribution (PSD) of a particle system.
The particle diameter of 63 μm (230 mesh) is measured using electroformed sieves and glass microsphere method.

Application

Used to validate and test particle size equipment.
Also used as a sieve calibration standard for:
  • characterizing electroformed sieves
  • investigating the effect of particle size on dissolution kinetics of magnesium leaching in titanium production

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • traceable to NIST and SRM standards
  • available as a neat sample

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors affecting the leaching mechanism of Mg bearing oxide impurities in Hydrogen Assisted Magnesiothermic Reduction (HAMR) of TiO2
Sarkar S, et al.
Chemosphere, 16, 100259-100259 (2020)
Sieve Calibration-a New Simple but High Precision Approach
Rideal GR, et al.
Particle & Particle Systems Characterization, 17(2), 77-82 (2000)

Related Content

Particle size conversion table from our Lab Basics Technical Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service