Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

49432

Supelco

Pyridine-3-sulfonyl chloride

for HPLC derivatization, LiChropur, ≥98.0% (GC)

Synonym(s):

3-Pyridinesulfonyl chloride, 3-Pyridylsulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClNO2S
Molecular Weight:
177.61
Beilstein:
120820
MDL number:
UNSPSC Code:
12352200

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥98.0% (GC)
97.0-103.0% (AT)

quality

LiChropur

technique(s)

HPLC: suitable

storage temp.

2-8°C

SMILES string

O=S(C1=CC=CN=C1)(Cl)=O

General description

Pyridine-3-sulfonyl chloride belongs to the class of substituted sulfonyl chlorides, which can be used as derivatization agents owing to their high proton affinity.

Application

Pyridine-3-sulfonyl chloride may be used as a derivatization agent to enhance the sensitivity of metabolites formed during the incubation of cytochrome P450 isoforms in aqueous medium, steroidal estrogens in biological samples, and bisphenols in beverages for their subsequent determination by high performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro cytochrome P450 activity: development and validation of a sensitive high performance liquid chromatography-tandem mass spectrometry method for the quantification of six probe metabolites after derivatization with pyridine-3-sulfonyl chloride in an aqueous environment
De Bock L, et al.
Journal of Chromatography A, 1218(6), 793-801 (2011)
Determination of bisphenols in beverages by mixed-mode solid-phase extraction and liquid chromatography coupled to tandem mass spectrometry
Regueiro J and Wenzl T
Journal of Chromatography A, 1422(1), 230-238 (2015)
Li Xu et al.
Analytical biochemistry, 375(1), 105-114 (2007-12-29)
Sulfonyl chlorides substituted with functional groups having high proton affinity can serve as derivatization reagents to enhance the sensitivity for steroidal estrogens in liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS). The most commonly used reagent for derivatization of estrogens
Chemical derivatization to enhance ionization of anabolic steroids in LC-MS for doping-control analysis.
Athanasiadou, I., et al.
Trends in Analytical Chemistry, 42, 137-156 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service