Skip to Content
MilliporeSigma
All Photos(1)

Documents

47564

Sigma-Aldrich

Fmoc-Tyr(PO3Bzl2)-OH

technical, ≥90% (HPLC), powder (amorphous)

Synonym(s):

Nα-Fmoc-O-[bis(benzyloxy)phosphoryl]-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H34NO8P
CAS Number:
Molecular Weight:
663.65
Beilstein:
4285612
MDL number:
UNSPSC Code:
12352200

grade

technical

Assay

≥90% (HPLC)

form

powder (amorphous)

optical activity

[α]20/D +33.0±2°, c = 1% in chloroform

storage temp.

−20°C

SMILES string

OC(=O)[C@H](Cc1ccc(OP(=O)(OCc2ccccc2)OCc3ccccc3)cc1)NC(=O)OCC4c5ccccc5-c6ccccc46

Other Notes

Building block for the synthesis of peptides with O-phospho-L-tyrosine

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.A. Kitas et al.
Helvetica Chimica Acta, 74, 1314-1314 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service