Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47442

Sigma-Aldrich

Fmoc-Asn-ONp

purum, ≥98.0% (calc. based on dry substance, C/N)

Synonym(s):

Fmoc-L-asparagine 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O7
CAS Number:
Molecular Weight:
475.45
Beilstein:
4617499
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥98.0% (calc. based on dry substance, C/N)

optical activity

[α]20/D −37±1°, c = 1% in DMF: AcOH (99:1)

mp

184-188 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

NC(=O)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(=O)Oc4ccc(cc4)[N+]([O-])=O

InChI

1S/C25H21N3O7/c26-23(29)13-22(24(30)35-16-11-9-15(10-12-16)28(32)33)27-25(31)34-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H2,26,29)(H,27,31)/t22-/m0/s1

InChI key

MPNFRQBESOZRJG-QFIPXVFZSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service