Skip to Content
MilliporeSigma
All Photos(1)

Documents

47390

Supelco

2-Fluorophenyl 2-nitrophenyl ether

Selectophore, ≥98.0%

Synonym(s):

FNDPE, 2-Fluoro-2′-nitrodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OC6H4NO2
CAS Number:
Molecular Weight:
233.20
Beilstein:
4457734
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥98.0% (GC)
≥98.0%

form

liquid

density

1.315 g/mL at 20 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Oc2ccccc2F

InChI

1S/C12H8FNO3/c13-9-5-1-3-7-11(9)17-12-8-4-2-6-10(12)14(15)16/h1-8H

InChI key

XVZDYNSMGNIZSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer in neutral carrier based polymer membrane ion-selective electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Sakamoto et al.
Analytical Chemistry, 59, 1513-1513 (1987)
O. Ryba et al.
Collection of Czechoslovak Chemical Communications, 49, 2371-2371 (1984)
I. Badr
Mikrochimica Acta, 149, 87-87 (2005)
Takashi Katsu et al.
Analytica chimica acta, 620(1-2), 50-54 (2008-06-19)
Although a previous study [S.S.M. Hassan, M.A. Ahmed, M.M. Saoudi, Anal. Chem. 57 (1985) 1126] had shown that a caffeine-sensitive electrode made with picrylsulfonate and 1-octanol as a cation-exchanger and a solvent mediator, respectively, had a wide working pH range
Lithium ion selective electrodes based on crown ethers for serum lithium assay.
K Kimura et al.
Analytical chemistry, 59(19), 2331-2334 (1987-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service