Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

46882

Supelco

Sulfanitran

VETRANAL®, analytical standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N4-Acetyl-N1-(4-nitrophenyl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3O5S
CAS Number:
Molecular Weight:
335.34
Beilstein:
2952955
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H13N3O5S/c1-10(18)15-11-4-8-14(9-5-11)23(21,22)16-12-2-6-13(7-3-12)17(19)20/h2-9,16H,1H3,(H,15,18)

InChI key

GWBPFRGXNGPPMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darko Ursic et al.
International journal of pharmaceutics, 381(2), 199-204 (2009-09-29)
Intestinal efflux transporters can significantly reduce the absorption of the drug after peroral application. In this work we studied secretion of glutathione conjugates triggered by glucose at the luminal side of the intestine. Glucose stimulated secretion of DNPSG, NEMSG and
Hehui Zheng et al.
Se pu = Chinese journal of chromatography, 25(2), 238-240 (2007-06-22)
A method for the determination of 12 sulfonamides (SAs) (sulfanilamide, sulfamonomethoxine, sulfacetamide, sulfamethoxazole, sulfadiazine, sulfisoxazole, sulfathiazole, sulfadi-methoxine, sulfamerazine, sulfaquinoxaline, sulfamethazine, sulfanitran) in cosmetics was developed by ultra performance liquid chromatography with photodiode array detector (UPLC-PDA). The chromatographic column used was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service