Skip to Content
MilliporeSigma
All Photos(1)

Documents

46830

Sigma-Aldrich

4-Fluorobenzyl chloride

purum, ≥98.0% (GC)

Synonym(s):

α-Chloro-4-fluorotoluene, 1-Chloromethyl-4-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2Cl
CAS Number:
Molecular Weight:
144.57
Beilstein:
742272
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.513 (lit.)
n20/D 1.513

bp

82 °C/26 mmHg (lit.)

density

1.207 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(CCl)cc1

InChI

1S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

IZXWCDITFDNEBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esakkiammal Sudha Esakkimuthu et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-18)
Lignin is an aromatic biopolymer derived from lignocellulosic biomass. Providing a comprehensive structural analysis of lignin is the primary motivation for the quantification of various functional groups, with a view to valorizing lignin in a wide range of applications. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service