Skip to Content
MilliporeSigma
All Photos(1)

Documents

46098

Sigma-Aldrich

Ethylene glycol diformate

≥98.0%

Synonym(s):

1,2-Diformyloxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOCH2CH2OCHO
CAS Number:
Molecular Weight:
118.09
Beilstein:
1746362
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0%

refractive index

n20/D 1.415

bp

174-178 °C (lit.)

density

1.226 g/mL at 20 °C (lit.)

SMILES string

O=COCCOC=O

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formate esters of 1,2-ethanediol: Major decomposition products of p-dioxane during storage.
D Jewett et al.
Bulletin of environmental contamination and toxicology, 25(1), 118-121 (1980-07-01)
Alexandra Chaskopoulou et al.
Journal of medical entomology, 46(6), 1400-1406 (2009-12-08)
Vapor toxicities of two prototype formate esters, ethylene glycol di-formate (EGDF) and heptyl formate, and the prototype heterobicyclic menthofuran were determined against laboratory-reared adult house flies, Musca domestica L. (Diptera: Muscidae). Toxicities were compared between house flies and other dipteran

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service