Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45968

Supelco

2,3-Dinitrotoluene

analytical standard

Synonym(s):

2,3-DNT, 2,4-DNT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
Beilstein:
2212428
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

59-61 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1cccc(c1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H6N2O4/c1-5-3-2-4-6(8(10)11)7(5)9(12)13/h2-4H,1H3

InChI key

DYSXLQBUUOPLBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dinitrotoluene is classified under the group of nitroaromatic compounds, which are organic nitro derivatives of analytical research interest. It is considered as a ubiquitous environmental contaminant and can adversely affect human health because of its mutagenic and/or carcinogenic nature.[1][2]

Application

2,3-Dinitrotoluene may be used as an analytical standard for the determination of the analyte in forensic samples, explosive-contaminated groundwater samples, industrial effluents and seawater samples by various chromatographic techniques.[3][4][5][1][6][7]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Hilmi et al.
Electrophoresis, 21(7), 1395-1404 (2000-05-29)
Amperometric detection at a bare gold electrode has been in-line coupled with capillary electrochromatography (CEC) for analysis of nitroaromatic and nitroamine explosives in contaminated soils and ground water. The CEC column packed with 3 microm C18 particles performed best using
Determination of dinitrotoluene isomers in sea water and industrial effluent by high-resolution electron-capture gas chromatography with a glass capillary column
Hashimoto A, et al.
Analyst, 105(1253), 787-793 (1980)
S C Reader et al.
Toxicology and applied pharmacology, 106(2), 287-294 (1990-11-01)
The present study was undertaken to evaluate the in vitro effects of four isomers of a known testicular toxicant, dinitroluene (DNT). Rat Sertoli or Sertoli-germ cell cocultures were treated, after 3 days in culture, with DNT isomers (0.01 to 100
Neera Singh et al.
Journal of hazardous materials, 173(1-3), 343-348 (2009-09-15)
The sorption of organic contaminants in soil is mainly attributed to the soil organic matter (SOM) content. However, recent studies have highlighted the fact that it is not the total carbon content of the organic matter, but its chemical structure
Igor A Buryakov
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 800(1-2), 75-82 (2003-12-31)
Description of a gas chromatograph designed for express analysis of explosives (2,4-dinitrotoluene, 2,4,6-trinitrotoluene, pentaerythritol tetranitrate), chemical warfare agents (mustard gas, lewisite, sarin) and drugs (heroin, cocaine hydrochloride, crack) is given. The devices comprises a multicapillary chromatographic column and an ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service