Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458600-M

Sigma-Aldrich

Ethanol (denatured)

UK IDA standard

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
grade:
denatured
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)

Product Name

Ethanol, denatured, (UK IDA standard)

grade

denatured

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

description

Source: fermented

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

availability

not available in Switzerland

refractive index

n20/D 1.3600 (lit.)

bp

78 °C (lit.)

mp

−114 °C (lit.)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • A highly efficient, selective, reversible and ultra-sensitive fluorescence "Turn-ON" chemosensor for aluminium ions by a novel Schiff base.: Demonstrates a novel application of ethanol in the synthesis of a fluorescent chemosensor, contributing to significant advancements in the detection of metal ions, which is crucial for environmental monitoring and industrial applications (Rana et al., 2024).
  • Illustration of the Intrinsic Mechanism of Reconstructed Cu Clusters for Enhanced CO(2) Electroreduction to Ethanol Production with Industrial Current Density.: This study provides insight into the catalytic processes involved in converting CO2 to ethanol, highlighting the pivotal role of ethanol in sustainable energy solutions (Wang G et al., 2024).
  • Br, O-Modified Cu(111) Interface Promotes CO(2) Reduction to Multicarbon Products.: Investigates the electrochemical reduction of CO2 over modified copper surfaces to produce ethanol and other multicarbon products, showcasing the integration of ethanol in chemical engineering and green chemistry (Cao R et al., 2024).
  • Catalytic Upgrading of Acetaldehyde to Acetoin using a Supported N-Heterocyclic Carbene Catalyst.: Discusses the catalytic transformation of acetaldehyde, a key intermediate in ethanol metabolism and industrial processes, to higher-value chemicals, underscoring the role of ethanol in synthetic pathways and industrial chemistry (Vorholt AJ et al., 2024).

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service