Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45779

Supelco

9,10-Dimethylanthracene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14
CAS Number:
Molecular Weight:
206.28
Beilstein:
1909028
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

182-184 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1c2ccccc2c(C)c3ccccc13

InChI

1S/C16H14/c1-11-13-7-3-5-9-15(13)12(2)16-10-6-4-8-14(11)16/h3-10H,1-2H3

InChI key

JTGMTYWYUZDRBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9,10-Dimethylanthracene belongs to the class of polynuclear aromatic hydrocarbons (PAHs), which are generally used as potential starting materials for the synthesis of a variety of industrial chemicals.[1]

Application

9,10-Dimethylanthracene may be used as an analytical standard for the determination of the analyte in coal and petroleum products as well as human biological samples by various chromatography techniques.[2][3][1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative high-performance liquid chromatographic determination of retinoids in human serum using on-line solid-phase extraction and column switching determination of 9-cis-retinoic acid, 13-cis-retinoic acid, all-trans-retinoic acid, 4-oxo-all-trans-retinoic acid and 4-oxo-13-cis-retinoic acid
Gundersen, TE, et al.
Journal of Chromatography. B, Biomedical Applications, 691(1), 43-58 (1997)
Analysis of polynuclear aromatic hydrocarbons in heavy products derived from coal and petroleum by high performance liquid chromatography
Zhang C, et al.
Journal of Chromatography A, 1167(2), 171-177 (2007)
Characterisation of middle-distillates by comprehensive two-dimensional gas chromatography (GC x GC): A powerful alternative for performing various standard analysis of middle-distillates
Vendeuvre C, et al.
Journal of Chromatography A, 1086(1-2), 21-28 (2005)
R K Saini et al.
Drug and chemical toxicology, 31(4), 459-471 (2008-10-14)
A single intratracheal instillation of 9,10-dimethyl benz(a)anthracene (DMBA) at 3 different doses of 5, 10, and 20 mg/kg body weight to Balb/c mice for 12 weeks had caused a significant incidence of pulmonary tumors along with inflammatory changes. The number
A smart polysaccharide/drug conjugate for photodynamic therapy.
So Young Park et al.
Angewandte Chemie (International ed. in English), 50(7), 1644-1647 (2011-02-11)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service