Skip to Content
MilliporeSigma
All Photos(1)

Documents

45763

Millipore

Ethyl acetate

suitable for HPLC, ≥99.8% (GC)

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.8% (GC)

autoignition temp.

801 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.05% water

evapn. residue

≤0.001%

refractive index

n20/D 1.3720 (lit.)
n20/D 1.372

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

solvent strength ε (Al2O3)

0.58

λ

1 cm path, H2O reference

UV absorption

λ: 255 nm Amax: 1.0
λ: 275 nm Amax: 0.1
λ: 300 nm Amax: 0.01

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junwei Liu et al.
Planta, 241(6), 1435-1451 (2015-02-27)
Strigolactone changes and cross talk with ABA unveil a picture of root-specific hormonal dynamics under stress. Strigolactones (SLs) are carotenoid-derived hormones influencing diverse aspects of development and communication with (micro)organisms, and proposed as mediators of environmental stimuli in resource allocation
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been
Maarit Neuvonen et al.
PloS one, 9(8), e103743-e103743 (2014-08-27)
Bacterial cholesterol oxidase is commonly used as an experimental tool to reduce cellular cholesterol content. That the treatment also generates the poorly degradable metabolite 4-cholesten-3-one (cholestenone) has received less attention. Here, we investigated the membrane partitioning of cholestenone using simulations
Tsukasa Horiyama et al.
PloS one, 9(9), e108642-e108642 (2014-09-27)
Escherichia coli produces the iron-chelating compound enterobactin to enable growth under iron-limiting conditions. After biosynthesis, enterobactin is released from the cell. However, the enterobactin export system is not fully understood. Previous studies have suggested that the outer membrane channel TolC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service