Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45590

Supelco

Monolinuron

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11ClN2O2
CAS Number:
Molecular Weight:
214.65
Beilstein:
2212523
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CON(C)C(=O)Nc1ccc(Cl)cc1

InChI

1S/C9H11ClN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)

InChI key

LKJPSUCKSLORMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mungi Kim et al.
The Science of the total environment, 757, 143815-143815 (2020-12-01)
Spatio-temporal distributions and bioaccumulation characteristics of freshwater cyanobacterial toxins, such as microcystins (MCs) in the Geum River Estuary, South Korea, were investigated during summer. Environmental multimedia samples (water, suspended particulate matter (SPM), and sediments) and tidal flat organisms (polychaetes, decapods
D Westphal et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 22(7), 521-528 (1984-07-01)
The biotransformation of radioactively labelled monolinuron (N-(4-chloro[U-14C]phenyl)-N'-methyl-N'-methoxyurea) was studied in the isolated perfused liver of the chicken. After a 4-hr perfusion, 83.1% of the added radioactivity was recovered, 56.6% in the perfusion medium and 26.5% in the liver and bile.
Methaemoglobinaemia due to monolinuron-not paraquat.
A T Proudfoot
British medical journal (Clinical research ed.), 285(6344), 812-812 (1982-09-18)
P B Casey et al.
Journal of toxicology. Clinical toxicology, 32(2), 185-189 (1994-01-01)
A 59-year-old man ingested a mouthful of Gramonol with suicidal intent and was admitted to a local hospital six and a half hours later. It was noted that he looked "ashen grey" and was centrally cyanosed. The methemoglobin and plasma
Studies of the arylhydroxylation of monochlorophenylureas in the isolated perfused rat liver.
D Westphal et al.
Food and cosmetics toxicology, 19(3), 341-345 (1981-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service