Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45576

Supelco

Methyl isothiocyanate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NCS
CAS Number:
Molecular Weight:
73.12
Beilstein:
605319
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

21 mmHg ( 20 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

117-118 °C (lit.)

mp

30-34 °C (lit.)

density

1.069 g/cm3 at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CN=C=S

InChI

1S/C2H3NS/c1-3-2-4/h1H3

InChI key

LGDSHSYDSCRFAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiji Nishiwaki et al.
Bioorganic & medicinal chemistry, 15(3), 1341-1345 (2006-11-28)
Tetrahydrobenzotriazines, a novel class of heterocyclic compounds, were synthesized. Examination of their biological activities resulted in the discovery that some of them possess potent nematocidal activity.
Luis O Ruzo
Pest management science, 62(2), 99-113 (2005-11-26)
Production and use of methyl bromide, a soil fumigant, are being restricted because of this chemical's deleterious effects on stratospheric ozone concentrations. Several products, some of which are currently used as soil fumigants, are being considered as possible replacements for
Y Zhang et al.
Journal of environmental quality, 34(5), 1566-1572 (2005-08-11)
Recent studies have observed enhanced degradation of methyl isothiocyanate (MITC) from repeated fumigation in agricultural soils. Little is known about fumigant degradation in forest and nursery soils. This study was conducted to determine degradation rates of MITC and chloropicrin (CP)
Michael L Dourson et al.
Regulatory toxicology and pharmacology : RTP, 58(2), 181-188 (2010-04-22)
The preplant fumigants, metam-sodium, metam-potassium, and dazomet undergo decomposition to the biocide methyl isothiocyanate (MITC) in moist soils. Since MITC vapor can migrate from its site of application, we developed an estimate of health protective concentrations for airborne exposures to
YongLin Ren et al.
Pest management science, 68(2), 194-201 (2011-07-23)
Methyl bromide is being phased out for use on stored commodities, as it is listed as an ozone-depleting substance, and phosphine is the fumigant widely used on grains. However, phosphine resistance occurs worldwide, and phosphine fumigation requires a long exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service