Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45519

Supelco

Genite

PESTANAL®, analytical standard

Synonym(s):

2,4-Dichlorophenyl benzenesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$71.90

$71.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$71.90

About This Item

Empirical Formula (Hill Notation):
C12H8Cl2O3S
CAS Number:
Molecular Weight:
303.16
Beilstein:
2944672
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$71.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(OS(=O)(=O)c2ccccc2)c(Cl)c1

InChI

1S/C12H8Cl2O3S/c13-9-6-7-12(11(14)8-9)17-18(15,16)10-4-2-1-3-5-10/h1-8H

InChI key

OZFAFGSSMRRTDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service