Skip to Content
MilliporeSigma
All Photos(1)

Documents

45356

Supelco

Bromoxynil-octanoate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17Br2NO2
CAS Number:
Molecular Weight:
403.11
Beilstein:
2756636
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCCCCC(=O)Oc1c(Br)cc(cc1Br)C#N

InChI

1S/C15H17Br2NO2/c1-2-3-4-5-6-7-14(19)20-15-12(16)8-11(10-18)9-13(15)17/h8-9H,2-7H2,1H3

InChI key

DQKWXTIYGWPGOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swen Follak et al.
Environmental pollution (Barking, Essex : 1987), 126(2), 139-146 (2003-08-21)
The present study was conducted to determine the effects of airborne herbicides on the photosynthesis and growth of non-target plants. Sunflowers at different growth stages were used as test plants and exposed for 24 h in a wind tunnel to
Tianming Cai et al.
Current microbiology, 63(2), 218-225 (2011-06-16)
Bromoxynil octanoate (BOO), the most widespread herbicide applied to maize, is potentially toxic to both animals and humans. In this article, a highly effective BOO-degrading bacterial strain, XB2, was isolated from the soil of a herbicide factory. The strain was
Xin-Gang Liu et al.
Journal of AOAC International, 92(3), 919-926 (2009-07-21)
In this paper, a simple, and rapid analysis of bromoxynil octanoate is reported for soil, corn leaves, and corn seeds by utilizing one-step liquid-liquid extraction and partitioning, followed by GC with electron capture (ECD) and mass spectrometry (MS) detection. The
Swen Follak et al.
Ecotoxicology (London, England), 14(5), 503-511 (2005-10-13)
Wind tunnel studies and a field study were conducted to investigate the impact of the airborne herbicide bromoxynil-octanoate on non-target plants. Sunflowers at the two-leaf stage were used as test plants and exposed for 24 h in a wind tunnel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service