Skip to Content
MilliporeSigma
All Photos(2)

Documents

45307

Supelco

Fensulfothion

PESTANAL®, analytical standard

Synonym(s):

Diethyl 4-(methylthio)phenyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17O4PS2
CAS Number:
Molecular Weight:
308.35
Beilstein:
2219515
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ccc(cc1)S(C)=O

InChI

1S/C11H17O4PS2/c1-4-13-16(17,14-5-2)15-10-6-8-11(9-7-10)18(3)12/h6-9H,4-5H2,1-3H3

InChI key

XDNBJTQLKCIJBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chandra et al.
Planta, 211(5), 736-742 (2000-11-23)
Because the H2O2 and O2- generated during a pathogen-triggered oxidative burst could either protect or destroy a besieged plant cell, their synthesis might be expected to be tightly regulated. We have examined the nature of this regulation as it is
J E Elliott et al.
Journal of wildlife diseases, 32(3), 486-491 (1996-07-01)
During the winter of 1990 in the Fraser Delta of British Columbia, Canada, nine birds of prey were found with symptoms of anticholinesterase poisoning. Immediate surgical removal of crop contents of three birds decreased mortality and recovery time. Chemical analysis
C Greenaway et al.
The Journal of emergency medicine, 14(3), 339-344 (1996-05-01)
We report a foodborne outbreak causing a cholinergic syndrome in three members of a family. The clinical presentation was characterized by nausea, vomiting, abdominal pain, and weakness. Physical examination revealed evidence of peripheral motor weakness and decreased level of consciousness
M M Sunil Paul et al.
Chemosphere, 91(3), 295-301 (2013-01-01)
Oxidative degradation of fensulfothion, a model organophosphorus compound, has been investigated by pulse radiolysis and H2O2/UV photolysis. A nearly complete transformation of fensulfothion was observed within 4min of irradiation. Very little Total Organic Carbon (TOC) reduction was obtained at this
Yi He et al.
Journal of chromatography. A, 1122(1-2), 7-12 (2006-05-24)
Continuous flow microextraction (CFME) combined with high-performance liquid chromatography-ultraviolet (HPLC-UV) detection has been applied to the analysis of five widely used pesticides, simazine, fensulfothion, etridiazole, mepronil and bensulide, present at trace levels in water samples. CFME employs a single organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service