Skip to Content
MilliporeSigma
All Photos(1)

Documents

45301

Supelco

Chlorbufam

PESTANAL®, analytical standard

Synonym(s):

1-Methyl-2-propynyl N-(3-chlorophenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10ClNO2
CAS Number:
Molecular Weight:
223.66
Beilstein:
4429630
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(OC(=O)Nc1cccc(Cl)c1)C#C

InChI

1S/C11H10ClNO2/c1-3-8(2)15-11(14)13-10-6-4-5-9(12)7-10/h1,4-8H,2H3,(H,13,14)

InChI key

ULBXWWGWDPVHAO-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Khafif et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 17(2), 153-170 (1982-01-01)
Kinetics of degradation of chlorbufam is determined in aqueous solution, in aqueous solution with humic acids, argilo-humic complexes, soil and sterile soil. The adsorption is also evaluated in this different fractions. The adsorption process has a protecting effect and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service