Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44970

Sigma-Aldrich

Iron(II) sulfate heptahydrate

puriss. p.a., ACS reagent

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

vapor pressure

14.6 mmHg ( 25 °C)

shelf life

~2 yr

impurities

≤0.001% total nitrogen (N)

pH

3.0-4.0 (25 °C, 50 mg/mL in H2O)

density

1.898 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤20 mg/kg
Cu: ≤10 mg/kg
Fe3+: ≤500 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter J Lukiw et al.
Journal of Alzheimer's disease & Parkinsonism, Suppl 2, 001-001 (2012-01-25)
The cellular generation of reactive oxygen species (ROS) has been implicated in contributing to the pathology of human neurological disorders including Alzheimer's disease (AD) and Parkinson's disease (PD). To further understand the triggering and participation of ROS-generating species to pro-inflammatory
Aileen I Pogue et al.
Journal of inorganic biochemistry, 105(11), 1434-1437 (2011-11-22)
Micro RNAs (miRNAs) constitute a unique class of small, non-coding ribonucleic acids (RNAs) that regulate gene expression at the post-transcriptional level. The presence of two inducible miRNAs, miRNA-125b and miRNA-146a, involved in respectively, astroglial cell proliferation and in the innate
Thermal analysis of iron (II) sulphate heptahydrate in air. V: Thermal decomposition of hydroxy and oxysulphates.
Swamy M and Prasad T.
Journal of Thermal Analysis and Calorimetry, 20(1), 107-114 (1981)
Thermal analysis of iron (II) sulphate heptahydrate in air: III. Thermal decomposition of intermediate hydrates.
Swami MSR and Prasad TP.
Journal of Thermal Analysis and Calorimetry, 19(2), 297-304 (1980)
Yanyan Wei et al.
Food chemistry, 138(2-3), 1952-1958 (2013-02-16)
The present study evaluated the effectiveness of germination and iron fortification on iron concentration and bioavailability of brown rice. Iron fortification during germination process with 0.05-2 g/L ferrous sulfate increased the iron concentration in germinated brown rice from 1.1 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service