Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44638

Supelco

3-(Methylthio)propionaldehyde

analytical standard

Synonym(s):

3-(Methylmercapto)propionaldehyde, 3-(Methylthio)propanal, Methional, NSC 15874

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CHO
CAS Number:
Molecular Weight:
104.17
Beilstein:
1739289
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

vapor density

>1 (vs air)

vapor pressure

760 mmHg ( 165 °C)

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n20/D 1.483 (lit.)

bp

165-166 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CSCCC=O

InChI

1S/C4H8OS/c1-6-4-2-3-5/h3H,2,4H2,1H3

InChI key

CLUWOWRTHNNBBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smaro Kokkinidou et al.
Journal of agricultural and food chemistry, 62(32), 8023-8033 (2014-07-30)
The application of phenolic compounds to suppress Maillard chemistry and off-flavor development in ultrahigh-termperature (UHT)-processed milk during processing and storage was investigated. Five phenolic compounds were examined for structure-reactivity relationships (catechin, genistein, daidzein, 1,2,3-trihydroxybenzene, and 1,3,5-trihydroxybenzene). The levels of key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service