Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44122

Sigma-Aldrich

8-Dodecanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt

for fluorescence, ≥95% (HPCE)

Synonym(s):

Trisodium 8-dodecanoyloxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29Na3O11S3
CAS Number:
Molecular Weight:
706.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥95% (HPCE)

mp

≥250 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 460 nm; λem 510 nm in 0.1 M Tris pH 8.0 (lipase)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].CCCCCCCCCCCC(=O)Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service