Skip to Content
MilliporeSigma
All Photos(1)

Documents

44042

Supelco

Silver ionophore VII

Selectophore, function tested

Synonym(s):

(3α,5β,12α)- 3-{[(2,2′-Bithiophen)-5-ylcarbonyl]oxy}-12-{{2-{[(diphenylamino)thioxomethyl]thio}acetyl}oxy}-N,N-dioctyl-cholan-24-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H88N2O5S4
CAS Number:
Molecular Weight:
1093.65
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

quality

function tested

storage temp.

2-8°C

SMILES string

CCCCCCCCN(CCCCCCCC)C(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3C[C@H](OC(=O)CSC(=S)N(c5ccccc5)c6ccccc6)[C@]12C)OC(=O)c7ccc(s7)-c8cccs8

InChI

1S/C64H88N2O5S4/c1-6-8-10-12-14-22-40-65(41-23-15-13-11-9-7-2)59(67)37-30-46(3)52-33-34-53-51-32-31-47-43-50(70-61(69)57-36-35-56(75-57)55-29-24-42-73-55)38-39-63(47,4)54(51)44-58(64(52,53)5)71-60(68)45-74-62(72)66(48-25-18-16-19-26-48)49-27-20-17-21-28-49/h16-21,24-29,35-36,42,46-47,50-54,58H,6-15,22-23,30-34,37-41,43-45H2,1-5H3/t46-,47-,50-,51+,52-,53+,54+,58+,63+,64-/m1/s1

InChI key

KKUPIKPCJBDFJI-CHUIGZLYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 7-deoxycholic amides or cholanes containing distinctive ion-recognizing groups at C3 and C12 and evaluation for ion-selective ionophores
On, J.H. et al.
Tetrahedron, 65, 1415-1423 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service