Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44030

Sigma-Aldrich

Dodecane

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
100 MG
500 MG
1 G


Fulfilment and delivery delayed



Select a Size

Change View
25 MG
100 MG
500 MG
1 G

About This Item

Linear Formula:
CH3(CH2)10CH3
CAS Number:
Molecular Weight:
170.33
Beilstein:
1697175
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:


Fulfilment and delivery delayed


vapor density

5.96 (vs air)

vapor pressure

1 mmHg ( 47.8 °C)

grade

technical

Assay

≥90% (GC)

autoignition temp.

401 °F

refractive index

n20/D 1.421 (lit.)
n20/D 1.421

bp

215-217 °C (lit.)

mp

−9.6 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F - Pensky-Martens closed cup

Flash Point(C)

70 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The novel extractants, diglycolamides, for the extraction of lanthanides and actinides in HNO3-n-dodecane system.
Sasaki Y, et al.
Solvent Extr. Ion Exch., 19(1), 91-103 (2001)
E J Walsh et al.
Biomedical microdevices, 8(1), 59-64 (2006-02-24)
The two-phase segmented flow approach to the processing and quantitative analysis of biological samples in microdevices offers significant advantages over the single-phase continuous flow methodology. Despite this, little is known about the compatibility of samples and reactants with segmenting fluids
Hydroisomerization and hydrocracking. 2. Product distributions from n-decane and n-dodecane.
Steijns M, et al.
Ind. Eng. Chem. Prod. Res. Dev., 20(4), 654-660 (1981)
Dorinde M M Kleinegris et al.
Enzyme and microbial technology, 48(3), 253-259 (2011-11-25)
During the in situ extraction of β-carotene from Dunaliella salina, the causal relationship between carotenoid extraction and cell death indicated that cell growth and cell death should be at equilibrium for a continuous in situ extraction process. In a flat-panel
I Solè et al.
Journal of colloid and interface science, 376(1), 133-139 (2012-04-07)
The influence of different dilution procedures on the properties of oil-in-water (O/W) nano-emulsions obtained by dilution of oil-in-water (O/W) and water-in-oil (W/O) microemulsions has been studied. The system water/SDS/cosurfactant/dodecane with either hexanol or pentanol as cosurfactant was chosen as model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service