Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43758

Supelco

(±)-1,2,4-Butanetriol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
106.12
Beilstein:
1733456
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

ASTM® D6584

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.473 (lit.)
n20/D 1.474

bp

190-191 °C/18 mmHg (lit.)

density

1.184 g/mL at 20 °C
1.19 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

OCCC(O)CO

InChI

1S/C4H10O3/c5-2-1-4(7)3-6/h4-7H,1-3H2

InChI key

ARXKVVRQIIOZGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Butanetriol (BT) is an important precursor utilized for the synthesis of the energetic material butanetriol trinitrate and several other chemicals. BT is also useful in the manufacture of plasticizers, polymers, cationic lipids, and pharmaceutical compounds.[1][2]

Application

1,2,4-Butanetriol may be used as an internal standard for the determination of glycerol,[3][4] and mono-, di-, and triglycerides[3] in vegetable oil methyl esters by capillary gas chromatography and gas chromatography coupled with flame ionization detector (GC-FID).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of glycerol, and mono-, di-and triglycerides in vegetable oil methyl esters by capillary gas chromatography
Plank C and Lorbeer E
Journal of Chromatography A, 697(1-2), 461-468 (1995)
Metabolic engineering of Arabidopsis for butanetriol production using bacterial genes
Abdel G, et al.
Metabolic engineering, 20(1), 109-120 (2013)
Direct bioconversion of d-xylose to 1, 2, 4-butanetriol in an engineered Escherichia coli
Valdehuesa K, et al.
Process Biochemistry (Oxford, United Kingdom), 49(1), 25-32 (2014)
Validation of a method for determination of free glycerol in biodiesel
Senila L, et al.
Studia Universitatis Babes-Bolyai, Chemia, 345(1-2), 461-468 (2016)
Ming Yan et al.
Journal of molecular biology, 427(8), 1765-1778 (2014-12-21)
The SAGA (Spt-Ada-Gcn5 acetyltransferase) complex performs multiple functions in transcription activation including deubiquitinating histone H2B, which is mediated by a subcomplex called the deubiquitinating module (DUBm). The yeast DUBm comprises a catalytic subunit, Ubp8, and three additional subunits, Sgf11, Sus1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service