Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43689

Supelco

4-Isopropylantipyrine

analytical standard

Synonym(s):

1,2-Dihydro-1,5-dimethyl-4-(1-methylethyl)-2-phenyl-3H-pyrazol-3-one, 1,2-Dihydro-4-isopropyl-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one, Isopropylphenazone, Propyphenazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O
CAS Number:
Molecular Weight:
230.31
Beilstein:
204533
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

O=C1C(C(C)C)=C(C)N(C)N1C2=CC=CC=C2

InChI

1S/C14H18N2O/c1-10(2)13-11(3)15(4)16(14(13)17)12-8-6-5-7-9-12/h5-10H,1-4H3

InChI key

PXWLVJLKJGVOKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Steiner et al.
Journal of visualized experiments : JoVE, (55)(55), doi:10-doi:10 (2011-09-29)
Hypersensitivity reactions against non-steroidal anti-inflammatory drugs (NSAIDs) like propyphenazone (PP) and diclofenac (DF) can manifest as Type I-like allergic reactions (1). In clinical practice, diagnosis of drug hypersensitivity is mainly performed by patient history, as skin testing is not reliable
Traugott J Scheytt et al.
Journal of contaminant hydrology, 83(1-2), 53-69 (2005-12-14)
Many pharmaceuticals pass the unsaturated zone before reaching an aquifer. Therefore, laboratory sand column transport experiments were conducted to study the transport behavior of carbamazepine, diclofenac, ibuprofen, and propyphenazone under unsaturated conditions. The test water was artificial sewage effluent to
Andreas Lemmerer et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(48), 13445-13460 (2011-11-15)
A co-crystal of two polymorphic active pharmaceutical ingredients (APIs), first reported and patented in 1937, has been prepared and thoroughly characterised, including crystal structure analysis. The existence of four crystal forms of one of the APIs, the sedative and hypnotic
Ghada M Hadad et al.
Journal of AOAC International, 90(4), 957-970 (2007-09-01)
Multivariate spectrophotometric calibration and liquid chromatography (LC) methods were used for the simultaneous determination of the active ingredients in 2 multicomponent mixtures containing chlorpheniramine maleate and phenylpropanolamine hydrochloride with ibuprofen and caffeine (mixture 1) or with propyphenazone (mixture 2). For
Acute inferior myocardial infarction with low atrial rhythm due to propyphenazone: Kounis syndrome.
Ahmet Akyel et al.
International journal of cardiology, 148(3), 352-353 (2010-06-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service