Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43581

Epoxide Hydrolase from Rhodotorula glutinis

BioChemika

Synonym(s):

Hydratase, epoxide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

product line

BioChemika

storage temp.

2-8°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service