Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43120

Sigma-Aldrich

Chloro(methyl)diphenylsilane

purum, ≥95.0% (GC)

Synonym(s):

Chloro-diphenyl-methylsilane, DPMSCl, Diphenylmethylchlorosilane, Methyldiphenylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(C6H5)2Cl
CAS Number:
Molecular Weight:
232.78
Beilstein:
2937445
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

vapor pressure

3 mmHg ( 125 °C)

grade

purum

Assay

≥95.0% (GC)

refractive index

n20/D 1.574 (lit.)

bp

295 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

C[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C13H13ClSi/c1-15(14,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11H,1H3

InChI key

OJZNZOXALZKPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protecting group for alcohols; The silyl ethers can be selectively cleaved with sodium azide in the presence of other silyl ethers (TBDMS, TBDPS); C-Silylation of ester enolates

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.E. Denmark et al.
The Journal of Organic Chemistry, 52, 165-165 (1987)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
G.L. Larson et al.
The Journal of Organic Chemistry, 53, 633-633 (1988)
S.J. Monger et al.
Journal of the Chemical Society. Chemical Communications, 381-381 (1989)
M Okamoto
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 112(6), 409-413 (1992-06-01)
The retention behavior and selectivity of 30 kinds of phenyl-modified porous glasses and silicas, prepared from solutions of phenyldimethylchlorosilane, diphenylmethylchlorosilane or triphenylchlorosilane in xylene, were studied by high-performance liquid chromatography using carbamazepine and diphenylhydantoin as model compounds. From the elemental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service