Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429546

Sigma-Aldrich

Propionaldehyde-2,4-dinitrophenylhydrazone

analytical standard, for environmental analysis

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$331.00

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$331.00

About This Item

Linear Formula:
C2H5CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard, for environmental analysis

Assay

99%

form

powder or crystals

mp

152-155 °C (lit.)

application(s)

diagnostic assay manufacturing
environmental
hematology
histology

storage temp.

room temp

SMILES string

CC\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H10N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h3-6,11H,2H2,1H3/b10-5+

InChI key

NFQHZOZOFGDSIN-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service