Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42365

Supelco

N-Methyldioctadecylamine

Selectophore, ≥99.0% (T)

Synonym(s):

N,N-Dioctadecylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)17]2NCH3
CAS Number:
Molecular Weight:
536.01
Beilstein:
1802318
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

Assay

≥99.0% (T)

mp

46-49 °C

SMILES string

CCCCCCCCCCCCCCCCCCN(C)CCCCCCCCCCCCCCCCCC

InChI

1S/C37H77N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38(3)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h4-37H2,1-3H3

InChI key

VFLWKHBYVIUAMP-UHFFFAOYSA-N

General description

N-Methyldioctadecylamine is a mono-alkyl tertiary amine.1 It is basically an hydrogen ionophore III.[1]
Visit our Sensor Applications portal to learn more.

Application

It may be used in preparing cation-sensitive PVC-based membranes for multisensory array used in measuring mineral water, soft drinks and beers.[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of quaternary ammonium salts methyl carbonate.
Geng, Tao, et al.
Fine Chemicals / ????, 6, 537-541 (2010)
All-solid-state electronic tongue and its application for beverage analysis.
Lvova, Larisa, et al.
Analytica Chimica Acta, 468, 303-314 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service