Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42198

Sigma-Aldrich

(S)-(+)-3,5-Dinitro-N-(1-phenylethyl)benzamide

for chiral derivatization

Synonym(s):

(S)-(+)-N-(3,5-Dinitrobenzoyl)-α-methylbenzylamine, (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCOC6H3(NO2)2
CAS Number:
Molecular Weight:
315.28
Beilstein:
3627249
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

derivatization grade ((chiral))
for chiral derivatization

optical activity

[α]20/D +47±2°, c = 0.9% in acetone

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

mp

158-161 °C (lit.)
158-161 °C

SMILES string

C[C@H](NC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O)c2ccccc2

InChI

1S/C15H13N3O5/c1-10(11-5-3-2-4-6-11)16-15(19)12-7-13(17(20)21)9-14(8-12)18(22)23/h2-10H,1H3,(H,16,19)/t10-/m0/s1

InChI key

ABEVDCGKLRIYRW-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service