Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42117

Supelco

Diisopropyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-bis(1-methylethyl) ester, NSC 15315, Phthalic acid diisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O4
CAS Number:
Molecular Weight:
250.29
Beilstein:
1972723
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.490 (lit.)
n20/D 1.490-1.500

density

1.063 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)OC(=O)c1ccccc1C(=O)OC(C)C

InChI

1S/C14H18O4/c1-9(2)17-13(15)11-7-5-6-8-12(11)14(16)18-10(3)4/h5-10H,1-4H3

InChI key

QWDBCIAVABMJPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisopropyl phthalate belongs to the class of phthalate esters, which are typically utilized as plasticizers to improve the flexibility of plastics in a broad range of household and consumer products. It is regarded as a potential endocrine disruptor.

Application

Diisopropyl phthalate may be used as an analytical standard for the determination of the analyte in meter dose inhalers (MDI) and children′s plastic toys by gas chromatography-tandem mass spectrometry (GC-MS/MS) technique.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phthalate esters from food-contacted materials by on-line microdialysis and liquid chromatography
Jen JF and Liu TC
Journal of Chromatography A, 1130(1), 28-33 (2006)
Analysis of 21 phthalate leachables in metered dose inhalers by gas chromatography tandem mass spectrometry
Ye X, et al.
Analytical Methods : Advancing Methods and Applications, 6(12), 4083-4089 (2014)
Optimization of multi-residue method for targeted screening and quantification of 216 harmful chemicals in plastic children?s toys by gas chromatography-tandem mass spectrometry analysis
Liu Y, et al.
Journal of Chromatography A, 1603(1), 311-326 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service