Skip to Content
MilliporeSigma
All Photos(1)

Documents

41753

Supelco

Spermine tetrahydrochloride

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N,N′-Bis(3-aminopropyl)-1,4-butanediamine tetrahydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H26N4 · 4HCl
CAS Number:
Molecular Weight:
348.18
Beilstein:
3911771
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

310-311 °C (dec.) (lit.)

format

neat

SMILES string

Cl.Cl.Cl.Cl.NCCCNCCCCNCCCN

InChI

1S/C10H26N4.4ClH/c11-5-3-9-13-7-1-2-8-14-10-4-6-12;;;;/h13-14H,1-12H2;4*1H

InChI key

XLDKUDAXZWHPFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Used to precipitate DNA from low salt aqueous buffers.

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA agonist/antagonist at the polyamine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service