Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41640-M

Sigma-Aldrich

Dimethyl sulfoxide

puriss. p.a., ACS reagent, ≥99.9% (GC)

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

GC/GC: suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.1% water

evapn. residue

≤0.005%

refractive index

n20/D 1.479 (lit.)
n20/D 1.479

bp

189 °C (lit.)

mp

16-19 °C (lit.)
18-20 °C

density

1.10 g/mL (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.5 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide (DMSO) is a polar solvent.[1] DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary, secondary, allylic, benzylic, hindered and bicyclic). Carbonyl compounds are formed as reaction products.[2] It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction, due to its high stability.[3]

Application


  • Ternary Phase-Field Simulation of Poly(vinylidene fluoride) Microporous Membrane Structures: Dimethyl sulfoxide (DMSO) is utilized in the formation of microporous membrane structures through phase separation techniques, enhancing polymer research for filtration and barrier applications (Zhang Z, et al., 2024).

  • Application of copper (I) selective ligands for PET imaging: DMSO is employed in the synthesis of copper (I) selective ligands for positron emission tomography (PET) imaging, illustrating its role in advancing diagnostic techniques in medical research (Tada T, et al., 2024).

  • Pillared graphene oxide frameworks for adsorption and separation: Research highlights the use of DMSO in exploring solvent interactions within graphene oxide frameworks, aiming to improve industrial processes for solvent recovery and environmental cleanup (Skarmoutsos I, et al., 2024).

  • A Luminescent Hybrid Bimetallic Halide Family with Solvent-Coordinated Rare Earth and Alkaline Earth Metals: This study utilizes DMSO in the development of new luminescent materials, showcasing its importance in the creation of advanced materials for lighting and display technologies (Lin J, et al., 2024).

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Other Notes

For information on dimethyl sulfoxide miscibility, please visit the following link:
Dimethyl Sulfoxide Miscibility/Immiscibility Table
Use as reagent in synthesis[4]

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgit Dreier et al.
Methods in molecular biology (Clifton, N.J.), 805, 261-286 (2011-11-19)
Ribosome display has proven to be a powerful in vitro selection and evolution method for generating high-affinity binders from libraries of folded proteins. It has been successfully applied to single-chain Fv fragments of antibodies and alternative scaffolds, such as Designed
Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)
A RAPID PERMETHYLATION OF GLYCOLIPID, AND POLYSACCHARIDE CATALYZED BY METHYLSULFINYL CARBANION IN DIMETHYL SULFOXIDE.
S HAKOMORI
Journal of biochemistry, 55, 205-208 (1964-02-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service