Skip to Content
MilliporeSigma
All Photos(1)

Documents

41611

Supelco

Cyclohexanecarboxylic acid

analytical standard

Synonym(s):

Hexahydrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CO2H
CAS Number:
Molecular Weight:
128.17
Beilstein:
970529
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)
97.5-102.5% (T)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.461 (lit.)

bp

232-233 °C (lit.)

mp

29-31 °C (lit.)

density

1.033 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(=O)C1CCCCC1

InChI

1S/C7H12O2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H,8,9)

InChI key

NZNMSOFKMUBTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexanecarboxylic acid, the saturated analog of benzoic acid, is categorized under the family of volatile organic compounds (VOCs). It is typically used as a flavoring ingredient.

Application

Cyclohexanecarboxylic acid may be used as an analytical standard for the determination of the analyte in wine and other alcoholic beverages, edible salts and high-salinity foods, and ethanolic extract of Centella asiatica by various analytical techniques.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of volatile organic acids in edible salt and high-salinity food by direct analysis in real time mass spectrometry
Fang P, et al.
International Journal of Mass Spectrometry, 444, 116166-116166 (2019)
Determination of 2-, 3-, 4-methylpentanoic and cyclohexanecarboxylic acids in wine: Development of a selective method based on solid phase extraction and gas chromatography-negative chemical ionization mass spectrometry and its application to different wines and alcoholic beverages
Gracia-Moreno E, et al.
Journal of Chromatography A, 1381, 210-218 (2015)
Essentials of organic chemistry: for students of pharmacy, medicinal chemistry and biological chemistry, 29, 328-334 (2013)
Phytochemistry, in vitro free radical scavenging, chelating and toxicity of Centela asiatica L.(Apiaceae) ethanolic leaf extract
Ghosh K and Indra N
International Journal of Pharmaceutical Sciences Review and Research, 29, 328-334 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service