Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41473

Millipore

Sucrose Fermentation Membrane

pinkish colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

Quality Level

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Klebsiella spp.
Proteus spp.
Serratia spp.
Streptococcus spp.
bacteria

General description

Sucrose Fermentation Membranes are substrate-carrying ID membranes used for rapid identification and differentiation of sucrose fermenting organisms through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on an acid indicator reaction in which specific sucrose fermenting organisms will exhibit yellow color.

Application

Sucrose Fermentation ID Membrane is used for rapid detection & confirmation of sucrose fermenting organisms in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Citrobacter freundii (8090) - PositiveEnterobacter aerogenes (13048) - PositiveEscherichia coli (25922)- NegativeKlebsiella pneumoniae (13883) - PositiveProteus vulgaris (13315) - PositiveSalmonella typhimurium (14028)- NegativeSalmonella typhi (6539)- NegativeSerratia marcescens (8100) - PositiveShigella flexneri (12022)- Negative

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service