Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41081

Supelco

Methyl 3-amino-4-methylthiophene-2-carboxylate

reference material

Synonym(s):

Articaine impurity I (PhEur), 3-Amino-4-methylthiophenecarboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
MDL number:
UNSPSC Code:
41116107

grade

reference material

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

85-88 °C (lit.)

SMILES string

COC(=O)c1scc(C)c1N

InChI

1S/C7H9NO2S/c1-4-3-11-6(5(4)8)7(9)10-2/h3H,8H2,1-2H3

InChI key

YICRPERKKBDRSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Jefferson et al.
Viruses, 12(1) (2020-01-08)
Sindbis virus (SINV) infection induces eIF2α phosphorylation, which leads to stress granule (SG) assembly. SINV infection also stimulates autophagy, which has an important role in controlling the innate immune response. The importance of autophagy to virus-induced translation arrest is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service