Skip to Content
MilliporeSigma
All Photos(1)

Documents

40998

Sigma-Aldrich

3-(N,N-Dimethyloctylammonio)propanesulfonate inner salt

purum, ≥95.0% (N)

Synonym(s):

Octyl sulfobetaine, SB3-8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
279.44
Beilstein:
3962489
MDL number:
UNSPSC Code:
12161900

description

zwitterionic

grade

purum

Assay

≥95.0% (N)

impurities

≤5% water

CMC

330 mM (20-25°C)

mp

171-174 °C
212-215 °C (lit.)

SMILES string

CCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C13H29NO3S/c1-4-5-6-7-8-9-11-14(2,3)12-10-13-18(15,16)17/h4-13H2,1-3H3

InChI key

QZRAABPTWGFNIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Zwitterionic detergent employed in capillary zone electrophoresis, CZE

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Senda et al.
Journal of High Resolution Chromatography, 15, 778-778 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service