Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40777

Supelco

Darutoside

analytical standard

Synonym(s):

(2R,4aS,4bR,7S,10aS)-7-[(1R)-1,2-Dihydroxyethyl]-1,2,3,4,4a,4b,5,6,7,9,10,10a-dodecahydro-1,1,4a,7-tetramethyl-2-phenanthrenyl-β-D-glucopyranoside, Darutigenol 3-β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44O8
CAS Number:
Molecular Weight:
484.62
Beilstein:
6542518
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@@H]2CC[C@]3(C)[C@@](CCC4=C[C@]([C@@H](O)CO)(C)CC[C@]43[H])([H])C2(C)C

InChI

1S/C26H44O8/c1-13-9-17-14(2)18(33-24-23(32)22(31)21(30)19(11-27)34-24)6-8-26(17,4)16-5-7-25(3,10-15(13)16)20(29)12-28/h10,13-14,16-24,27-32H,5-9,11-12H2,1-4H3/t13?,14?,16-,17+,18?,19-,20+,21-,22+,23-,24?,25+,26-/m1/s1

InChI key

OJRJAOXVGPMJAP-XOQGPCJVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service