Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40775

Supelco

Nano Silica Gel Adamant on TLC Plates

glass plates

Synonym(s):

Silica Gel Nano Adamant on TLC Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110504

form

glass plates

feature

binder Organic
fluorescent indicator: no

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

5 cm × 5 cm

layer thickness

200 μm

pore size

60 Å medium pore diameter

Looking for similar products? Visit Product Comparison Guide

General description

Nanotechnology led to the production of nanosized synthetic crystalline form silica. Nanosilica can be synthesized using reverse microemulsion, flame synthesis and sol-gel methods.[1]

Application

Nano Silica Gel Adamant on TLC Plates may be used in HPTLC and TLC.

Legal Information

Adamant is a trademark of Macherey-Nagel GmbH & Co., KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of silica nanoparticles by sol-gel: size-dependent properties, surface modification, and applications in silica-polymer nanocomposites-a review
Rahman, Ismail Ab, and Vejayakumaran Padavettan.
Journal of Nanomaterials, 8, 1-15 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service