Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40724

Supelco

Norfluoxetine hydrochloride

analytical standard

Synonym(s):

(±)-γ-(4-Trifluoromethylphenoxy)benzenepropanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16F3NO · HCl
CAS Number:
Molecular Weight:
331.76
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].NCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2

InChI

1S/C16H16F3NO.ClH/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;/h1-9,15H,10-11,20H2;1H

InChI key

GMTWWEPBGGXBTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevgi Başalan Över et al.
Arhiv za higijenu rada i toksikologiju, 71(3), 211-222 (2020-10-20)
The aim of this study was to investigate the oxidative and apoptotic potential of fluoxetine, a widely used antidepressant in Turkey and the world, and of its metabolite norfluoxetine on a model non-target organism, Daphnia magna to see how exposure
Yin Yao Dong et al.
Science (New York, N.Y.), 347(6227), 1256-1259 (2015-03-15)
TREK-2 (KCNK10/K2P10), a two-pore domain potassium (K2P) channel, is gated by multiple stimuli such as stretch, fatty acids, and pH and by several drugs. However, the mechanisms that control channel gating are unclear. Here we present crystal structures of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service