Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40721

Supelco

2-Propanethiol

analytical standard

Synonym(s):

Isopropyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHSH
CAS Number:
Molecular Weight:
76.16
Beilstein:
605260
EC Number:
MDL number:
UNSPSC Code:
12352105
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor density

2.6 (vs air)

vapor pressure

455 mmHg ( 37.8 °C)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.425-1.427
n20/D 1.426 (lit.)

bp

57-60 °C (lit.)

mp

−131 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)S

InChI

1S/C3H8S/c1-3(2)4/h3-4H,1-2H3

InChI key

KJRCEJOSASVSRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanethiol is a sulfur compound, which belongs to the family of volatile organic compounds (VOCs). It is a potential aroma ingredient of a variety of foods and beverages.

Application

2-Propanethiol may be used as an analytical standard for the determination of the analyte in beer, food products, fruits, and industrial effluents by various chromatography techniques.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-23.8 °F - closed cup

Flash Point(C)

-31 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The perfume of carabao mangoes (Mangifera indica L.)
Naef R, et al.
European Food Research and Technology, 222(5-6), 554-558 (2006)
New procedures for control of industrial effluents treatment processes
Boczkaj G, et al.
Industrial & Engineering Chemistry Research, 53(4), 1503-1514 (2014)
Isothermal Kovats retention indices of sulfur compounds on a poly (5\% diphenyl-95\% dimethylsiloxane) stationary phase
Miller KE and Bruno TJ
Journal of Chromatography A, 1007(1-2), 117-125 (2003)
Determination of sulphur compounds in beer using headspace solid-phase microextraction and gas chromatographic analysis with pulsed flame photometric detection
Hill PG and Smith RM
Journal of Chromatography A, 872(1-2), 203-213 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service