Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40545

Millipore

CaCO3 Agar

suitable for microbiology

Synonym(s):

Agar with calcium carbonate, Custer′s Chalk Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

sterility

non-sterile

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
CaCO3 (fine granulated), 5 g/L
glucose, 50 g/L
yeast extract, 5 g/L

application(s)

clinical testing
food and beverages

microbiology

suitability

nonselective and differential for Candida spp.
nonselective and differential for Pichia spp.
nonselective and differential for Saccharomyces spp.
nonselective and differential for Zygosaccharomyces spp.
nonselective and differential for molds (General Media)
nonselective and differential for yeasts (General Media)

Application

CaCO3 Agar is a medium for the differentiation of microorganisms based on the production of acid from glucose. Kurtzman and Fell (1998) recommended the formulation for the yeast identification.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service